CAS RN 7789-69-7
IUPAC Name:
pentabromo-λ⁵-phosphane
Synonyms:
phosphorus pentabromide
Molecular Weight (g/mol):
430.49
Molecular Formula:
Br5P
InChi Key:
QRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILES:
BrP(Br)(Br)(Br)Br