accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFCa

CAS RN 7789-75-5

IUPAC Name: calcium difluoride
Synonyms: fluorite|calcium fluoride|calcium difluoride|CaF2|kalziumfluorid|kalziumdifluorid
Molecular Weight (g/mol): 78.08
Molecular Formula: CaF2
InChi Key: WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES: [F-].[F-].[Ca++]
Quantity 
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (30)
Percent Purity 
  • (8)
  • (2)
  • (9)
  • (3)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (6)
  • (3)
  • (16)
Grade 
  • (3)
  • (3)
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (28)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (30)
  • (8)
  • (2)
  • (9)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (16)
  • (3)
  • (3)
  • (7)
  • (2)
  • (2)