accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-99-3

IUPAC Name: 2-methylpentyl acetate
Synonyms: 1-pentanol, 2-methyl-, acetate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: UZTVUHTZGGZFCI-UHFFFAOYNA-N
SMILES: CCCC(C)COC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results