accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 779-03-3

NH2

CAS RN 779-03-3

IUPAC Name: anthracen-9-amine
Synonyms: 9-aminoanthracene 10-anthracylamine
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: LHNICELDCMPPDE-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)