CAS RN 779-30-6
CAS RN 779-30-6
IUPAC Name:
N-(2-oxo-2H-chromen-3-yl)acetamide
Synonyms:
N-(2-oxochromen-3-yl)acetamide
Molecular Weight (g/mol):
203.2
Molecular Formula:
C11H9NO3
InChi Key:
XJYLCCJIDYSFMB-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC2=CC=CC=C2OC1=O
1
–
1
of
1
results
1
3-Acetamidocoumarin 98.0+%, TCI America™
CAS: 779-30-6 Molecular Formula: C11H9NO3 Molecular Weight (g/mol): 203.20 MDL Number: MFCD00075556 InChI Key: XJYLCCJIDYSFMB-UHFFFAOYSA-N PubChem CID: 136620 IUPAC Name: N-(2-oxo-2H-chromen-3-yl)acetamide SMILES: CC(=O)NC1=CC2=CC=CC=C2OC1=O