accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7790-33-2

IUPAC Name: manganese(2+) diiodide
Synonyms: manganese(II) iodide
Molecular Weight (g/mol): 308.75
Molecular Formula: I2Mn
InChi Key: QWYFOIJABGVEFP-UHFFFAOYSA-L
SMILES: [Mn++].[I-].[I-]
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)