accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7790-44-5

IUPAC Name: antimony(3+) triiodide
Synonyms: antimony triiodide
Molecular Weight (g/mol): 502.47
Molecular Formula: I3Sb
InChi Key: KWQLUUQBTAXYCB-UHFFFAOYSA-K
SMILES: [Sb+3].[I-].[I-].[I-]
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)