accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaCaOPOOOPOOO

CAS RN 7790-76-3

IUPAC Name: dicalcium (phosphonatooxy)phosphonate
Synonyms: calcium pyrophosphate ca2P2O7 diphosphoric acid, calcium salt (2:1) calcium diphosphate
Molecular Weight (g/mol): 254.1
Molecular Formula: Ca2O7P2
InChi Key: JUNWLZAGQLJVLR-UHFFFAOYSA-J
SMILES: [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)