accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OCdOSOOO

CAS RN 7790-84-3

IUPAC Name: cadmium(2+) hydrate sulfate
Synonyms: cadmium(2+) hydrate sulfate
Molecular Weight (g/mol): 226.49
Molecular Formula: CdH2O5S
InChi Key: VRFRVKRTIMIRNG-UHFFFAOYSA-L
SMILES: O.[Cd++].[O-]S([O-])(=O)=O
Grade 
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
Physical Form 
  • (3)
  • (5)
  • (7)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (5)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (3)
  • (1)
  • (7)
  • (6)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (5)
  • (7)
  • (3)
  • (3)
  • (1)
  • (5)
  • (4)
  • (5)
  • (2)
  • (3)