accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSeClO

CAS RN 7791-23-3

IUPAC Name: selenooyl dichloride
Synonyms: selenium oxychloride selenium oxydichloride
Molecular Weight (g/mol): 165.87
Molecular Formula: Cl2OSe
InChi Key: LIRMPIXWGWLNND-UHFFFAOYSA-N
SMILES: Cl[Se](Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results