accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77943-39-6

IUPAC Name: 4-methyl-5-phenyl-1,3-oxazolidin-2-one
Synonyms: 4-methyl-5-phenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: PPIBJOQGAJBQDF-UHFFFAOYNA-N
SMILES: CC1NC(=O)OC1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)