CAS RN 77989-15-2
CAS RN 77989-15-2
IUPAC Name:
2-(2-bromophenyl)-4,6-diphenyl-1,3,5-triazine
Synonyms:
2-(2-bromophenyl)-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol):
388.27
Molecular Formula:
C21H14BrN3
InChi Key:
MSTJGWCHJCZPEQ-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC=C1C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
CAS: 77989-15-2 Molecular Formula: C21H14BrN3 Molecular Weight (g/mol): 388.268 MDL Number: MFCD29047055 InChI Key: MSTJGWCHJCZPEQ-UHFFFAOYSA-N PubChem CID: 12689834 IUPAC Name: 2-(2-bromophenyl)-4,6-diphenyl-1,3,5-triazine SMILES: C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=CC=C3Br)C4=CC=CC=C4