accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77992-44-0

IUPAC Name: 5-bromo-2-hydrazinylpyridine
Synonyms: 5-bromo-2-hydrazinylpyridine
Molecular Weight (g/mol): 188.03
Molecular Formula: C5H6BrN3
InChi Key: QYQLEYTXFMOLEI-UHFFFAOYSA-N
SMILES: NNC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results