accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiH3COH3COH3C

CAS RN 78-07-9

IUPAC Name: triethoxy(ethyl)silane
Synonyms: ethyl triethoxysilane
Molecular Weight (g/mol): 192.33
Molecular Formula: C8H20O3Si
InChi Key: DENFJSAFJTVPJR-UHFFFAOYSA-N
SMILES: CCO[Si](CC)(OCC)OCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)