CAS RN 78-10-4
Tetraethyl orthosilicate, 98%
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.33 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethyl orthosilicate, 98%, AcroSeal™
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.33 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethoxysilane, 98%
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Ethyl Silicate, Condensed, Spectrum™ Chemical
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.33 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethoxysilane, 99+%
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethoxysilane, 99.9%
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethoxysilane, 99+%
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethoxysilane, 99.999+% (metals basis)
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Tetraethyl Orthosilicate 97.0+%, TCI America™
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 MDL Number: MFCD00009062 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
Sigma Aldrich Tetraethyl orthosilicate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetraethyl orthosilicate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 5-Sulfosalicylic Acid Dihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetraethyl orthosilicate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetraethyl Orthosilicate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Ethyl Silicate, Reag, MP Biomedicals
CAS: 78-10-4 Molecular Formula: C8H20O4Si Molecular Weight (g/mol): 208.329 InChI Key: BOTDANWDWHJENH-UHFFFAOYSA-N Synonym: tetraethyl orthosilicate,tetraethoxysilane,teos,ethyl silicate,silicon ethoxide,silicon tetraethoxide,silane, tetraethoxy,dynasil a,tetraethoxysilicon,ethyl orthosilicate PubChem CID: 6517 IUPAC Name: tetraethyl silicate SMILES: CCO[Si](OCC)(OCC)OCC
- 1
- 2