accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-26-2

IUPAC Name: 2-methyl-2-propylpropane-1,3-diol
Synonyms: 2-methyl-2-propylpropane-1,3-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: JVZZUPJFERSVRN-UHFFFAOYSA-N
SMILES: CCCC(C)(CO)CO
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)