accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-54-6

IUPAC Name: 1-[2-(1-hydroxycyclohexyl)ethynyl]cyclohexan-1-ol
Synonyms: 1-[2-(1-hydroxycyclohexyl)ethynyl]cyclohexan-1-ol
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: YMAIXWWPLZFJLA-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C#CC1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results