accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-63-7

IUPAC Name: 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
Synonyms: varox
Molecular Weight (g/mol): 290.44
Molecular Formula: C16H34O4
InChi Key: DMWVYCCGCQPJEA-UHFFFAOYSA-N
SMILES: CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)