accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-69-3

IUPAC Name: 3,7-dimethyloctan-3-ol
Synonyms: 3-octanol, 3,7-dimethyl-
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: DLHQZZUEERVIGQ-UHFFFAOYNA-N
SMILES: CCC(C)(O)CCCC(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)