accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-74-0

IUPAC Name: 1,1,2-tribromoethane
Synonyms: ethane, 1,1,2-tribromo-
Molecular Weight (g/mol): 266.76
Molecular Formula: C2H3Br3
InChi Key: QUMDOMSJJIFTCA-UHFFFAOYSA-N
SMILES: BrCC(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results