accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 78-98-8

IUPAC Name: 2-oxopropanal
Synonyms: pyruvoyl group aldehyde, pyruvic acetalformaldehyde propanolone 1-ketopropionaldehyde pyroracemic aldehyde α-ketopropionic aldehyde acetylformyl
Molecular Weight (g/mol): 72.06
Molecular Formula: C3H4O2
InChi Key: AIJULSRZWUXGPQ-UHFFFAOYSA-N
SMILES: CC(=O)C=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)
Boiling Point 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)