accessibility menu, dialog, popup

UserName

Viewing profile as user:
OVOONH4

CAS RN 7803-55-6

IUPAC Name: vanadiumoylol amine
Synonyms: ammonium vanadate
Molecular Weight (g/mol): 116.98
Molecular Formula: H4NO3V
InChi Key: YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES: N.O[V](=O)=O
Grade 
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (6)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
Physical Form 
  • (5)
  • (3)
  • (8)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (7)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (4)
  • (2)
  • (3)
  • (7)
  • (2)
  • (6)
  • (4)
  • (14)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
  • (5)
  • (3)
  • (8)
  • (6)
  • (2)
  • (1)
  • (3)
  • (7)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)