CAS RN 78032-05-0
CAS RN 78032-05-0
IUPAC Name:
6-methylphthalazine
Synonyms:
6-methylphthalazine
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
NPOVJYAUMRFAQD-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C=NN=CC2=C1
1
–
1
of
1
results
1
6-Methylphthalazine 98.0+%, TCI America™
CAS: 78032-05-0 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.18 MDL Number: MFCD03844800 InChI Key: NPOVJYAUMRFAQD-UHFFFAOYSA-N PubChem CID: 12679508 IUPAC Name: 6-methylphthalazine SMILES: CC1=CC=C2C=NN=CC2=C1