accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78078-50-9

IUPAC Name: 2-[2-(bromomagnesio)ethyl]-1,3-dioxane
Synonyms: 2-[2-(bromomagnesio)ethyl]-1,3-dioxane
Molecular Weight (g/mol): 219.36
Molecular Formula: C6H11BrMgO2
InChi Key: JYNXRXBIEHSSLR-UHFFFAOYSA-M
SMILES: Br[Mg]CCC1OCCCO1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)