accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 780800-85-3

IUPAC Name: 5-phenylpyridine-2-carbaldehyde
Synonyms: 5-phenylpyridine-2-carbaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: IMMPVOLAPKOFFB-UHFFFAOYSA-N
SMILES: O=CC1=NC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results