accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78095-19-9

IUPAC Name: 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide
Synonyms: 8-OH-dpat hydrobromide
Molecular Weight (g/mol): 328.29
Molecular Formula: C16H26BrNO
InChi Key: BATPBOZTBNNDLN-UHFFFAOYNA-N
SMILES: Br.CCCN(CCC)C1CCC2=C(C1)C(O)=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results