accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 781-17-9

IUPAC Name: 4,5,9,10-tetrahydropyrene
Synonyms: 4,5,9,10-tetrahydropyrene
Molecular Weight (g/mol): 206.29
Molecular Formula: C16H14
InChi Key: XDFUNRTWHPWCKO-UHFFFAOYSA-N
SMILES: C1CC2=C3C(CCC4=C3C1=CC=C4)=CC=C2
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results