CAS RN 78110-38-0
Thermo Scientific Chemicals Aztreonam
CAS: 78110-38-0 Molecular Formula: C13H17N5O8S2 Molecular Weight (g/mol): 435.426 InChI Key: WZPBZJONDBGPKJ-PSGLRMFWSA-N PubChem CID: 56842327 IUPAC Name: 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid SMILES: CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
Aztreonam, 95.7%, MP Biomedicals™
CAS: 78110-38-0 Molecular Formula: C13H17N5O8S2 Molecular Weight (g/mol): 435.426 InChI Key: WZPBZJONDBGPKJ-PSGLRMFWSA-N Synonym: Azthreonam,Azactam PubChem CID: 56842327 IUPAC Name: 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid SMILES: CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
Aztreonam MP Biomedicals
CAS: 78110-38-0 Molecular Formula: C13H17N5O8S2 Molecular Weight (g/mol): 435.426 InChI Key: WZPBZJONDBGPKJ-PSGLRMFWSA-N Synonym: trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid,2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid PubChem CID: 56842327 IUPAC Name: 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid SMILES: CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
Aztreonam, MilliporeSigma™ Supelco™
Aztreonam is a synthetic monocyclic β-lactum, which can be isolated from Chromobacterium violaceum. It may find clinical applications as a potent antimicrobial agent, due to its bactericidal action.
Aztreonam
CAS: 78110-38-0 Molecular Formula: C13H17N5O8S2 Molecular Weight (g/mol): 435.426 MDL Number: MFCD00072145 InChI Key: WZPBZJONDBGPKJ-PSGLRMFWSA-N PubChem CID: 56842327 IUPAC Name: 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid SMILES: CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N
Aztreonam Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Aztreonam 98.0+%, TCI America™
CAS: 78110-38-0 Molecular Formula: C13H17N5O8S2 Molecular Weight (g/mol): 435.426 MDL Number: MFCD00072145 InChI Key: WZPBZJONDBGPKJ-PSGLRMFWSA-N Synonym: trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid,2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid PubChem CID: 56842327 IUPAC Name: 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid SMILES: CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N