accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78119-82-1

IUPAC Name: 6-hydroxynaphthalene-2-carbaldehyde
Synonyms: 6-hydroxynaphthalene-2-carbaldehyde
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: PRYNJOJHKYNLIS-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=C(C=O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results