accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78151-58-3

CH3NOONOH3CO

CAS RN 78151-58-3

IUPAC Name: 7,18-dioctyl-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: 7,18-dioctyl-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Weight (g/mol): 614.79
Molecular Formula: C40H42N2O4
InChi Key: YFGMQDNQVFJKTR-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C(=O)C2=CC=C3C4=CC=C5C(=O)N(CCCCCCCC)C(=O)C6=CC=C(C7=CC=C(C1=O)C2=C37)C4=C56
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)