accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFOOO

CAS RN 78153-79-4

IUPAC Name: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-fluorooxane
Synonyms: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-fluorooxane
Molecular Weight (g/mol): 542.65
Molecular Formula: C34H35FO5
InChi Key: QNXIKNZDQVSBCO-UHFFFAOYNA-N
SMILES: FC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results