CAS RN 78162-58-0
IUPAC Name:
N,N-dimethyl-1H-imidazole-1-sulfonamide
Synonyms:
N,N-dimethylimidazole-1-sulfonamide
Molecular Weight (g/mol):
175.21
Molecular Formula:
C5H9N3O2S
InChi Key:
YRRWNBMOJMMXQY-UHFFFAOYSA-N
SMILES:
CN(C)S(=O)(=O)N1C=CN=C1
1
–
1
of
1
results
1
N,N-Dimethyl-1H-imidazole-1-sulfonamide 98.0+%, TCI America™
CAS: 78162-58-0 Molecular Formula: C5H9N3O2S Molecular Weight (g/mol): 175.206 MDL Number: MFCD00955713 InChI Key: YRRWNBMOJMMXQY-UHFFFAOYSA-N Synonym: 1-(Dimethylsulfamoyl)-1H-imidazole PubChem CID: 395751 IUPAC Name: N,N-dimethylimidazole-1-sulfonamide SMILES: CN(C)S(=O)(=O)N1C=CN=C1