accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 781654-31-7

IUPAC Name: 3-bromo-4,5-dimethoxybenzonitrile
Synonyms: 3-bromo-4,5-dimethoxybenzonitrile
Molecular Weight (g/mol): 242.07
Molecular Formula: C9H8BrNO2
InChi Key: PTCVIEQABWPBTA-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(Br)=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results