accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONSOOCl

CAS RN 782501-25-1

IUPAC Name: tert-butyl 4-(chlorosulfonyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(chlorosulfonyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 283.77
Molecular Formula: C10H18ClNO4S
InChi Key: VJAHMDQRVLEOFG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)S(Cl)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)