accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78291-99-3

IUPAC Name: 7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),5,8,10-tetraen-11-amine
Synonyms: 7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),5,8,10-tetraen-11-amine
Molecular Weight (g/mol): 191.25
Molecular Formula: C9H9N3S
InChi Key: HJCGGAYYNMECPO-UHFFFAOYSA-N
SMILES: NC1=CC=C2SC3=NCCN3C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results