accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 78322-96-0

IUPAC Name: 4-(pyrimidin-2-yl)benzonitrile
Synonyms: 4-(pyrimidin-2-yl)benzonitrile
Molecular Weight (g/mol): 181.2
Molecular Formula: C11H7N3
InChi Key: WGCKVXHOTDDNOA-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results