accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 78358-86-8

IUPAC Name: 1-(2-bromoethyl)-1H-pyrrole
Synonyms: 1-(2-bromoethyl)pyrrole
Molecular Weight (g/mol): 174.04
Molecular Formula: C6H8BrN
InChi Key: QBAVHEZVBGASER-UHFFFAOYSA-N
SMILES: BrCCN1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)