accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78385-26-9

IUPAC Name: 3-(bromomethyl)-3-methyloxetane
Synonyms: 3-(bromomethyl)-3-methyloxetane
Molecular Weight (g/mol): 165.03
Molecular Formula: C5H9BrO
InChi Key: MGBZKWOJRYGRTO-UHFFFAOYSA-N
SMILES: CC1(CBr)COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results