accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 784-38-3

IUPAC Name: 4-chloro-2-(2-fluorobenzoyl)aniline
Synonyms: 4-chloro-2-(2-fluorobenzoyl)aniline
Molecular Weight (g/mol): 249.67
Molecular Formula: C13H9ClFNO
InChi Key: GTGMXPIQRQSORU-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results