accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONH2Cl

CAS RN 78433-88-2

IUPAC Name: 2-(2,6-dichlorophenyl)acetamide
Synonyms: 2-(2,6-dichlorophenyl)acetamide
Molecular Weight (g/mol): 204.05
Molecular Formula: C8H7Cl2NO
InChi Key: KEPLFZWNVMCJLI-UHFFFAOYSA-N
SMILES: NC(=O)CC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results