accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78440-76-3

IUPAC Name: 3-(methoxymethyl)-1H-indole
Synonyms: 3-(methoxymethyl)-1H-indole
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: RJWLFBCRNXZXTJ-UHFFFAOYSA-N
SMILES: COCC1=CNC2=CC=CC=C12
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results