accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOI

CAS RN 78489-65-3

IUPAC Name: ethyl 2-iodo-2-phenylacetate
Synonyms: ethyl 2-iodo-2-phenylacetate
Molecular Weight (g/mol): 290.1
Molecular Formula: C10H11IO2
InChi Key: GFYUBPKXVMWINH-UHFFFAOYNA-N
SMILES: CCOC(=O)C(I)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results