accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78546-66-4

IUPAC Name: N-[4-(2-hydroxyphenyl)-1,3-thiazol-2-yl]acetamide
Synonyms: N-[4-(2-hydroxyphenyl)-1,3-thiazol-2-yl]acetamide
Molecular Weight (g/mol): 234.27
Molecular Formula: C11H10N2O2S
InChi Key: RCVLPHKAZAQDGZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(=CS1)C1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results