accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78573-45-2

IUPAC Name: 3-[3-(trifluoromethyl)phenyl]propan-1-ol
Synonyms: 3-[3-(trifluoromethyl)phenyl]propan-1-ol
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H11F3O
InChi Key: QWXKQVIMGVVIBX-UHFFFAOYSA-N
SMILES: OCCCC1=CC(=CC=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results