accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3SSCl

CAS RN 786-19-6

IUPAC Name: O,O-diethyl ({[(4-chlorophenyl)sulfanyl]methyl}sulfanyl)phosphonothioate
Synonyms: carbophenothion 卡波硫磷 carbophenthion
Molecular Weight (g/mol): 342.85
Molecular Formula: C11H16ClO2PS3
InChi Key: VEDTXTNSFWUXGQ-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCSC1=CC=C(Cl)C=C1
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results