accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78600-31-4

IUPAC Name: 2-bromo-N,N-diphenylaniline
Synonyms: 2-bromo-N,N-diphenylaniline
Molecular Weight (g/mol): 324.22
Molecular Formula: C18H14BrN
InChi Key: YPIANBZIVBPMJS-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results