accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78600-33-6

IUPAC Name: 3-bromo-N,N-diphenylaniline
Synonyms: 3-bromo-N,N-diphenylaniline
Molecular Weight (g/mol): 324.22
Molecular Formula: C18H14BrN
InChi Key: YDXLVFKTOSKBKT-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results