man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 78617-12-6

H3COOOHOHOHHO

CAS RN 78617-12-6

IUPAC Name: 2-(heptyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: heptyl glucoside �-D-galactopyranoside, heptyl
Molecular Weight (g/mol): 278.35
Molecular Formula: C13H26O6
InChi Key: NIDYWHLDTIVRJT-UHFFFAOYNA-N
SMILES: CCCCCCCOC1OC(CO)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results