CAS RN 78686-77-8
IUPAC Name:
methyl 5-bromo-6-chloropyridine-3-carboxylate
Synonyms:
methyl 5-bromo-6-chloropyridine-3-carboxylate
Molecular Weight (g/mol):
250.48
Molecular Formula:
C7H5BrClNO2
InChi Key:
WINGWVOUOFMOJQ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=C(Cl)C(Br)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 5-bromo-6-chloronicotinate, 98%
Methyl 5-bromo-6-chloronicotinate, 98%
CAS: 78686-77-8 Molecular Formula: C7H5BrClNO2 Molecular Weight (g/mol): 250.476 MDL Number: MFCD03844855 InChI Key: WINGWVOUOFMOJQ-UHFFFAOYSA-N PubChem CID: 2764313 IUPAC Name: methyl 5-bromo-6-chloropyridine-3-carboxylate SMILES: COC(=O)C1=CC(=C(N=C1)Cl)Br