accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78712-43-3

IUPAC Name: hydrogen (2E)-3-{4-[(1H-imidazol-1-yl)methyl]phenyl}prop-2-enoic acid chloride
Synonyms: hydrogen ozagrel chloride
Molecular Weight (g/mol): 264.71
Molecular Formula: C13H13ClN2O2
InChi Key: CWKFWBJJNNPGAM-IPZCTEOASA-N
SMILES: [H+].[Cl-].OC(=O)\C=C\C1=CC=C(CN2C=CN=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results